Assay Details
| Gene Symbol: QKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053446.1 | XP_017908935.1 | 3 - 4 | 504 | 64 | |
| XM_018053447.1 | XP_017908936.1 | 3 - 4 | 504 | 64 | ||
| XM_018053448.1 | XP_017908937.1 | 3 - 4 | 504 | 64 | ||
| XM_018053449.1 | XP_017908938.1 | 3 - 4 | 531 | 64 | ||
| XM_018053450.1 | XP_017908939.1 | 3 - 4 | 532 | 64 | ||
| XM_018053451.1 | XP_017908940.1 | 3 - 4 | 531 | 64 | ||
| GenBank mRNA | JO377413.2 | - | 3 - 4 | 634 | 64 | |
More Information
Set Membership: |
|

QKI
Genomic Map