Assay Details
| Gene Symbol: ARPC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055757.1 | XP_017911246.1 | 2 - 3 | 361 | 95 | |
| GenBank mRNA | JO510467.2 | - | 2 - 3 | 718 | 95 | |
| JQ308784.1 | - | 2 - 3 | 291 | 95 | ||
More Information
Set Membership: |
|

ARPC5L
Genomic Map