Assay Details
| Gene Symbol: PPIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042255.1 | XP_017897744.1 | 3 - 4 | 438 | 64 | |
| GenBank mRNA | JO411140.2 | - | 3 - 4 | 422 | 64 | |
More Information
Set Membership: |
|

PPIF
Genomic Map