Assay Details
| Gene Symbol: HRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692705.3 | XP_005692762.2 | 3 - 4 | 2277 | 64 | |
| XM_013971550.2 | XP_013827004.2 | 3 - 4 | 2277 | 64 | ||
| GenBank mRNA | JP350892.2 | - | 4 - 5 | 1971 | 64 | |
| JP350894.2 | - | 4 - 5 | 1986 | 64 | ||
More Information
Set Membership: |
|

HRC
Genomic Map