Assay Details
| Gene Symbol: LOC102191423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965702.2 | XP_013821156.1 | 8 - 9 | 854 | 108 | |
| XM_013965703.2 | XP_013821157.1 | 8 - 9 | 959 | 108 | ||
| XM_013965704.2 | XP_013821158.1 | 10 - 11 | 1156 | 108 | ||
| XM_013965705.2 | XP_013821159.1 | 9 - 10 | 1076 | 108 | ||
| XM_018050132.1 | XP_017905621.1 | 8 - 9 | 959 | 108 | ||
| XM_018050133.1 | XP_017905622.1 | 8 - 9 | 959 | 108 | ||
| XM_018050134.1 | XP_017905623.1 | 8 - 9 | 959 | 108 | ||
| GenBank mRNA | JT714845.1 | - | 10 - 11 | 1079 | 108 | |
More Information
Set Membership: |
|

LOC102191423
Genomic Map