Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677434.3 | XP_005677491.2 | 6 - 7 | 552 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TDRD10| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677434.3 | XP_005677491.2 | 6 - 7 | 552 | 65 | |
Set Membership: |
|