Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681129.2 | XP_005681186.1 | 1 - 2 | 348 | 50 | |
| Gene Symbol: PHETA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048834.1 | XP_017904323.1 | 3 - 4 | 3231 | 50 | |
| XM_018048835.1 | XP_017904324.1 | 3 - 4 | 3187 | 50 | ||
| XM_018048836.1 | XP_017904325.1 | 3 - 4 | 3154 | 50 | ||
| XM_018048837.1 | XP_017904326.1 | 3 - 4 | 3125 | 50 | ||
| XM_018048838.1 | XP_017904327.1 | 3 - 4 | 3206 | 50 | ||
| XM_018048839.1 | XP_017904328.1 | 3 - 4 | 3185 | 50 | ||
More Information
Set Membership: |
|

SMDT1
Genomic Map