Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681129.2 | XP_005681186.1 | 2 - 3 | 487 | 94 | |
| Gene Symbol: PHETA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048834.1 | XP_017904323.1 | 4 - 5 | 3370 | 94 | |
| XM_018048835.1 | XP_017904324.1 | 4 - 5 | 3326 | 94 | ||
| XM_018048836.1 | XP_017904325.1 | 4 - 5 | 3293 | 94 | ||
| XM_018048837.1 | XP_017904326.1 | 4 - 5 | 3264 | 94 | ||
| XM_018048838.1 | XP_017904327.1 | 4 - 5 | 3345 | 94 | ||
| XM_018048839.1 | XP_017904328.1 | 4 - 5 | 3324 | 94 | ||
| XM_018048840.1 | XP_017904329.1 | 4 - 5 | 3299 | 94 | ||
More Information
Set Membership: |
|

SMDT1
Genomic Map