Assay Details
| Gene Symbol: ARL6IP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970605.2 | XP_013826059.1 | 3 - 4 | 557 | 64 | |
| XM_018061343.1 | XP_017916832.1 | 3 - 4 | 641 | 64 | ||
| XM_018061344.1 | XP_017916833.1 | 3 - 4 | 616 | 64 | ||
| XM_018061345.1 | XP_017916834.1 | 3 - 4 | 640 | 64 | ||
| XM_018061346.1 | XP_017916835.1 | 3 - 4 | 615 | 64 | ||
More Information
Set Membership: |
|

ARL6IP4
Genomic Map