Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699602.3 | XP_005699659.2 | 2 - 3 | 472 | 68 | |
| XM_005699604.3 | XP_005699661.2 | 3 - 4 | 587 | 68 | ||
| XM_005699606.3 | XP_005699663.2 | 2 - 3 | 504 | 68 | ||
| GenBank mRNA | KA346503.1 | - | 3 - 4 | 1280 | 68 | |
More Information
Set Membership: |
|

PUS3
Genomic Map