Assay Details
| Gene Symbol: NOL4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057704.1 | XP_017913193.1 | 7 - 8 | 1482 | 61 | |
| XM_018057705.1 | XP_017913194.1 | 7 - 8 | 1482 | 61 | ||
| XM_018057706.1 | XP_017913195.1 | 4 - 5 | 822 | 61 | ||
More Information
Set Membership: |
|

NOL4L
Genomic Map