Assay Details
| Gene Symbol: CCM2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057688.1 | XP_017913177.1 | 9 - 10 | 1438 | 57 | |
| XM_018057689.1 | XP_017913178.1 | 9 - 10 | 1435 | 57 | ||
More Information
Set Membership: |
|

CCM2L
Genomic Map