Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058144.1 | XP_017913633.1 | 4 - 5 | 1152 | 61 | |
| XM_018058145.1 | XP_017913634.1 | 4 - 5 | 1152 | 61 | ||
| GenBank mRNA | JO411730.2 | - | 5 - 6 | 755 | 61 | |
| JO428894.2 | - | 5 - 6 | 1633 | 61 | ||
More Information
Set Membership: |
|

DEPTOR
Genomic Map