Assay Details
| Gene Symbol: SLC30A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060161.1 | XP_017915650.1 | 3 - 4 | 1024 | 56 | |
| GenBank mRNA | JO413414.2 | - | 3 - 4 | 1017 | 56 | |
More Information
Set Membership: |
|

SLC30A10
Genomic Map