Assay Details
| Gene Symbol: POLR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682900.3 | XP_005682957.2 | 2 - 3 | 415 | 59 | |
| XM_018050470.1 | XP_017905959.1 | 2 - 3 | 409 | 59 | ||
| GenBank mRNA | JO349118.2 | - | 5 - 6 | 514 | 59 | |
More Information
Set Membership: |
|

POLR2E
Genomic Map