Assay Details
| Gene Symbol: KLHL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684660.3 | XP_005684717.1 | 4 - 5 | 753 | 55 | |
| XM_013966475.2 | XP_013821929.1 | 4 - 5 | 753 | 55 | ||
More Information
Set Membership: |
|

KLHL32
Genomic Map