Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695793.3 | XP_005695850.1 | 5 - 6 | 886 | 74 | |
| GenBank mRNA | JO434173.2 | - | 5 - 6 | 882 | 74 | |
| JT674038.1 | - | 5 - 6 | 871 | 74 | ||
More Information
Set Membership: |
|

DNASE1L3
Genomic Map