Assay Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062816.1 | XP_017918305.1 | 5 - 6 | 766 | 75 | |
| XM_018062817.1 | XP_017918306.1 | 5 - 6 | 627 | 75 | ||
More Information
Set Membership: |
|

IL4I1
Genomic Map