Assay Details
| Gene Symbol: ACSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061873.1 | XP_017917362.1 | 2 - 3 | 758 | 62 | |
| XM_018061874.1 | XP_017917363.1 | 2 - 3 | 758 | 62 | ||
| XM_018061875.1 | XP_017917364.1 | 2 - 3 | 758 | 62 | ||
| XM_018061876.1 | XP_017917365.1 | 2 - 3 | 758 | 62 | ||
| GenBank mRNA | JO434435.2 | - | 2 - 3 | 734 | 62 | |
More Information
Set Membership: |
|

ACSF3
Genomic Map