Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695244.3 | XP_005695301.1 | 3 - 4 | 1457 | 69 | |
| GenBank mRNA | JO434465.2 | - | 3 - 4 | 684 | 69 | |
| JT673412.1 | - | 7 - 8 | 826 | 69 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map