Assay Details
| Gene Symbol: LOC102187646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963513.2 | XP_013818967.2 | 1 - 2 | 157 | 71 | |
| GenBank mRNA | JO446723.2 | - | 4 - 5 | 635 | 71 | |
| JT665813.1 | - | 1 - 2 | 173 | 71 | ||
More Information
Set Membership: |
|

LOC102187646
Genomic Map