Assay Details
| Gene Symbol: LOC102187646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963513.2 | XP_013818967.2 | 6 - 7 | 750 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102187646| Gene Symbol: LOC102187646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963513.2 | XP_013818967.2 | 6 - 7 | 750 | 64 | |
Set Membership: |
|