Assay Details
| Gene Symbol: LOC102187646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963513.2 | XP_013818967.2 | 8 - 9 | 914 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102187646| Gene Symbol: LOC102187646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963513.2 | XP_013818967.2 | 8 - 9 | 914 | 65 | |
Set Membership: |
|