Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968695.2 | XP_013824149.2 | 3 - 4 | 806 | 69 | |
| XM_018064978.1 | XP_017920467.1 | 3 - 4 | 817 | 69 | ||
| GenBank mRNA | JO444393.2 | - | 1 - 2 | 186 | 69 | |
More Information
Set Membership: |
|

HOXD3
Genomic Map