Assay Details
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061664.1 | XP_017917153.1 | 9 - 10 | 823 | 70 | |
| XM_018061665.1 | XP_017917154.1 | 8 - 9 | 814 | 70 | ||
| XM_018061666.1 | XP_017917155.1 | 9 - 10 | 824 | 70 | ||
| XM_018061667.1 | XP_017917156.1 | 9 - 10 | 823 | 70 | ||
| XM_018061668.1 | XP_017917157.1 | 8 - 9 | 814 | 70 | ||
More Information
Set Membership: |
|

MTSS2
Genomic Map