Assay Details
| Gene Symbol: LOC102190263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065523.1 | XP_017921012.1 | 16 - 17 | 2002 | 68 | |
| XM_018065524.1 | XP_017921013.1 | 15 - 16 | 2602 | 68 | ||
| XM_018065527.1 | XP_017921016.1 | 16 - 17 | 2003 | 68 | ||
More Information
Set Membership: |
|

LOC102190263
Genomic Map