Assay Details
| Gene Symbol: LOC102190263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065523.1 | XP_017921012.1 | 23 - 24 | 3053 | 100 | |
| XM_018065524.1 | XP_017921013.1 | 22 - 23 | 3653 | 100 | ||
| XM_018065527.1 | XP_017921016.1 | 24 - 25 | 3255 | 100 | ||
More Information
Set Membership: |
|

LOC102190263
Genomic Map