Assay Details
| Gene Symbol: LOC102190263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065523.1 | XP_017921012.1 | 4 - 5 | 438 | 55 | |
| XM_018065524.1 | XP_017921013.1 | 3 - 4 | 1038 | 55 | ||
| XM_018065527.1 | XP_017921016.1 | 4 - 5 | 439 | 55 | ||
More Information
Set Membership: |
|

LOC102190263
Genomic Map