Assay Details
| Gene Symbol: LOC102190263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065523.1 | XP_017921012.1 | 8 - 9 | 964 | 90 | |
| XM_018065524.1 | XP_017921013.1 | 7 - 8 | 1564 | 90 | ||
| XM_018065527.1 | XP_017921016.1 | 8 - 9 | 965 | 90 | ||
More Information
Set Membership: |
|

LOC102190263
Genomic Map