Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040934.1 | XP_017896423.1 | 2 - 3 | 690 | 87 | |
| XM_018040935.1 | XP_017896424.1 | 2 - 3 | 690 | 87 | ||
| XM_018040936.1 | XP_017896425.1 | 2 - 3 | 694 | 87 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map