Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042908.1 | XP_017898397.1 | 4 - 5 | 610 | 57 | |
| GenBank mRNA | JO445253.2 | - | 2 - 3 | 329 | 57 | |
| JO524738.2 | - | 1 - 2 | 395 | 57 | ||
| JO579419.1 | - | 4 - 5 | 610 | 57 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map