Assay Details
| Gene Symbol: MRPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056868.1 | XP_017912357.1 | 2 - 3 | 318 | 67 | |
| GenBank mRNA | JO321503.2 | - | 1 - 2 | 580 | 67 | |
| JO368184.2 | - | 1 - 2 | 552 | 67 | ||
| JO392739.2 | - | 2 - 3 | 393 | 67 | ||
| JO397130.2 | - | 2 - 3 | 256 | 67 | ||
| JO517199.2 | - | 1 - 2 | 558 | 67 | ||
| JO535558.2 | - | 2 - 3 | 312 | 67 | ||
| JO569260.2 | - | 1 - 2 | 527 | 67 | ||
| JO572130.2 | - | 1 - 2 | 527 | 67 | ||
| KA336944.1 | - | 1 - 2 | 639 | 67 | ||
More Information
Set Membership: |
|

MRPS6
Genomic Map