Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679054.3 | XP_005679111.1 | 2 - 3 | 604 | 101 | |
| XM_005679055.3 | XP_005679112.1 | 3 - 4 | 685 | 101 | ||
| GenBank mRNA | JO443242.2 | - | 3 - 4 | 682 | 101 | |
More Information
Set Membership: |
|

CCDC126
Genomic Map