Assay Details
| Gene Symbol: ANAPC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691210.3 | XP_005691267.3 | 2 - 3 | 825 | 97 | |
| XM_018061572.1 | XP_017917061.1 | 2 - 3 | 397 | 97 | ||
| XM_018061573.1 | XP_017917062.1 | 2 - 3 | 234 | 97 | ||
| GenBank mRNA | KA308255.1 | - | 3 - 4 | 522 | 97 | |
More Information
Set Membership: |
|

ANAPC10
Genomic Map