Assay Details
| Gene Symbol: LOC102181148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063150.1 | XP_017918639.1 | 4 - 5 | 3470 | 61 | |
| GenBank mRNA | JO635200.2 | - | 2 - 3 | 266 | 61 | |
More Information
Set Membership: |
|

LOC102181148
Genomic Map