Assay Details
| Gene Symbol: ZNF875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063403.1 | XP_017918892.1 | 2 - 3 | 462 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF875| Gene Symbol: ZNF875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063403.1 | XP_017918892.1 | 2 - 3 | 462 | 69 | |
Set Membership: |
|