Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065666.1 | XP_017921155.1 | 3 - 4 | 499 | 74 | |
| XM_018065667.1 | XP_017921156.1 | 3 - 4 | 500 | 74 | ||
| GenBank mRNA | JO613346.2 | - | 4 - 5 | 2030 | 74 | |
More Information
Set Membership: |
|

OTULIN
Genomic Map