Assay Details
| Gene Symbol: IGSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046367.1 | XP_017901856.1 | 3 - 4 | 1103 | 58 | |
| XM_018046368.1 | XP_017901857.1 | 4 - 5 | 1012 | 58 | ||
More Information
Set Membership: |
|

IGSF8
Genomic Map