Assay Details
| Gene Symbol: ATPAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043806.1 | XP_017899295.1 | 8 - 9 | 864 | 65 | |
| XM_018043811.1 | XP_017899300.1 | 7 - 8 | 813 | 65 | ||
More Information
Set Membership: |
|

ATPAF1
Genomic Map