Assay Details
| Gene Symbol: C8H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684108.3 | XP_005684165.1 | 3 - 4 | 1042 | 80 | |
| XM_018052418.1 | XP_017907907.1 | 3 - 4 | 507 | 80 | ||
More Information
Set Membership: |
|

C8H9orf64
Genomic Map