Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692364.3 | XP_005692421.1 | 3 - 4 | 1070 | 89 | |
| XM_005692365.3 | XP_005692422.1 | 3 - 4 | 1070 | 89 | ||
| XM_013971160.2 | XP_013826614.1 | 3 - 4 | 1194 | 89 | ||
| GenBank mRNA | JT678663.1 | - | 2 - 3 | 125 | 89 | |
| KA311967.1 | - | 3 - 4 | 434 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005692365.3 transcripts.
Set Membership: |
|

ZNF527
Genomic Map