Assay Details
| Gene Symbol: RDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063106.1 | XP_017918595.1 | 4 - 5 | 566 | 58 | |
| XM_018063107.1 | XP_017918596.1 | 4 - 5 | 566 | 58 | ||
| XM_018063108.1 | XP_017918597.1 | 3 - 4 | 337 | 58 | ||
More Information
Set Membership: |
|

RDH13
Genomic Map