Assay Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694751.3 | XP_005694808.2 | 12 - 13 | 1930 | 54 | |
| XM_013972794.2 | XP_013828248.2 | 11 - 12 | 1865 | 54 | ||
| XM_013972795.2 | XP_013828249.2 | 11 - 12 | 1865 | 54 | ||
More Information
Set Membership: |
|

C6
Genomic Map