Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045950.1 | XP_017901439.1 | 2 - 3 | 232 | 60 | |
| GenBank mRNA | JO616996.2 | - | 6 - 7 | 774 | 60 | |
| JT686656.1 | - | 2 - 3 | 184 | 60 | ||
More Information
Set Membership: |
|

TSPAN2
Genomic Map