Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056779.1 | XP_017912268.1 | 4 - 5 | 354 | 71 | |
| XM_018056780.1 | XP_017912269.1 | 4 - 5 | 352 | 71 | ||
| GenBank mRNA | JO617289.2 | - | 2 - 3 | 544 | 71 | |
| JT741291.1 | - | 4 - 5 | 245 | 71 | ||
More Information
Set Membership: |
|

DNAJC15
Genomic Map