Assay Details
| Gene Symbol: LOC102180880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693171.3 | XP_005693228.1 | 2 - 3 | 426 | 88 | |
| GenBank mRNA | JO325964.2 | - | 2 - 3 | 286 | 88 | |
| JO613021.2 | - | 2 - 3 | 640 | 88 | ||
More Information
Set Membership: |
|

LOC102180880
Genomic Map