Assay Details
| Gene Symbol: APOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694476.3 | XP_005694533.1 | 3 - 4 | 400 | 62 | |
| XM_005694477.3 | XP_005694534.1 | 2 - 3 | 223 | 62 | ||
More Information
Set Membership: |
|

APOH
Genomic Map