Assay Details
| Gene Symbol: MRI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051217.1 | XP_017906706.1 | 5 - 6 | 1035 | 71 | |
| GenBank mRNA | JO416040.2 | - | 1 - 2 | 286 | 71 | |
| JO646483.2 | - | 1 - 2 | 239 | 71 | ||
More Information
Set Membership: |
|

MRI1
Genomic Map