Assay Details
| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050995.1 | XP_017906484.1 | 4 - 5 | 599 | 87 | |
| GenBank mRNA | EU244432.1 | - | 4 - 5 | 509 | 87 | |
| JI861618.1 | - | 4 - 5 | 509 | 87 | ||
| JO392878.2 | - | 1 - 2 | 219 | 87 | ||
| JQ417407.1 | - | 4 - 5 | 509 | 87 | ||
More Information
Set Membership: |
|

RAB11B
Genomic Map