Assay Details
| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058697.1 | XP_017914186.1 | 9 - 10 | 2742 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARP10| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058697.1 | XP_017914186.1 | 9 - 10 | 2742 | 70 | |
Set Membership: |
|